Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252377.1 | NP_001239306.1 | 2 | 277 | ||
NM_014595.2 | NP_055410.1 | 2 | 277 | |||
NR_045513.1 | 2 | 277 | ||||
XM_011524700.2 | XP_011523002.2 | 2 | 447 | |||
GenBank mRNA | AF154829.2 | - | 2 | 264 | ||
AK000419.1 | - | 2 | 264 | |||
BC008183.1 | - | 2 | 225 | 4183256 | ||
BC017454.1 | - | 2 | 242 | 4647090 | ||
BC022334.1 | - | 2 | 224 | 4703567 | ||
BG034032.1 | - | 2 | 164 | |||
BQ431723.1 | - | 2 | 277 | |||
EU794640.1 | - | 2 | 264 |