Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303047.1 | NP_001289976.1 | 4 | 687 | ||
NM_001303048.1 | NP_001289977.1 | 3 | 330 | |||
NM_001303049.1 | NP_001289978.1 | 3 | 337 | |||
NM_006471.3 | NP_006462.1 | 3 | 604 | |||
GenBank mRNA | AK222569.1 | - | 3 | 280 | ||
AK291145.1 | - | 3 | 300 | |||
AW958488.1 | - | 3 | 330 | |||
BC016372.1 | - | 3 | 279 | 4293343 | ||
BC031972.1 | - | 3 | 604 | 4836883 | ||
BC032748.1 | - | 3 | 275 | 5563221 | ||
BM561281.1 | - | 4 | 404 | |||
CD702288.1 | - | 3 | 360 | |||
D82059.1 | - | 3 | 270 | |||
DQ185044.1 | - | 3 | 322 | |||
EU794621.1 | - | 3 | 604 | |||
X54304.1 | - | 3 | 323 |