Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012265.1 | NP_036397.1 | 6 | 1343 | ||
XM_006724224.3 | XP_006724287.1 | 5 | 1096 | |||
XM_011530107.2 | XP_011528409.1 | 6 | 1273 | |||
XM_017028749.1 | XP_016884238.1 | 3 | 1859 | |||
XM_017028750.1 | XP_016884239.1 | 5 | 1661 | |||
GenBank mRNA | AL050346.1 | - | 6 | 1343 | ||
BC002705.2 | - | 4 | 994 | 3627843 | ||
BC018872.2 | - | 4 | 994 | |||
CR456411.1 | - | 5 | 1049 | |||
CR457382.1 | - | 4 | 971 |