Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 6 | 1954 | ||
NM_001301141.1 | NP_001288070.1 | 6 | 1965 | |||
NM_001320042.1 | NP_001306971.1 | 5 | 3041 | |||
NM_014301.4 | NP_055116.1 | 6 | 1964 | |||
NM_213595.3 | NP_998760.1 | 5 | 1868 | |||
NR_135127.1 | 7 | 2064 | ||||
XM_011538100.2 | XP_011536402.1 | 6 | 3137 | |||
XM_011538101.2 | XP_011536403.1 | 4 | 1789 | |||
GenBank mRNA | AK000574.1 | - | 5 | 711 | ||
AK022758.1 | - | 4 | 3465 | |||
AY009127.1 | - | 6 | 819 | |||
AY009128.1 | - | 5 | 723 | |||
BC011906.2 | - | 5 | 724 | 4136262 | ||
BC061903.1 | - | 5 | 717 | 4183674 | ||
BG621612.1 | - | 335 | ||||
EU329002.1 | - | 7 | 921 | |||
U47101.1 | - | 4 | 571 |