Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014448.3 | NP_055263.2 | 11, 12 | 1873 | ||
XM_017001049.1 | XP_016856538.1 | 11, 12 | 2474 | |||
XM_017001050.1 | XP_016856539.1 | 11, 12 | 1679 | |||
XM_017001051.1 | XP_016856540.1 | 11, 12 | 1884 | |||
GenBank mRNA | AK095984.1 | - | 11, 12 | 1780 | ||
AK296150.1 | - | 6, 7 | 771 | |||
BC002681.1 | - | 10, 11 | 1160 | 3613623 | ||
BC051838.1 | - | 9, 10 | 1180 | 6191377 | ||
BT007270.1 | - | 6, 7 | 761 | |||
D89016.1 | - | 8, 9 | 1188 |