Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252377.1 | NP_001239306.1 | 3 | 408 | ||
NM_014595.2 | NP_055410.1 | 3 | 408 | |||
NR_045513.1 | 3 | 384 | ||||
GenBank mRNA | AF154829.2 | - | 3 | 395 | ||
AK000419.1 | - | 3 | 395 | |||
BC008183.1 | - | 3 | 332 | 4183256 | ||
BC017454.1 | - | 3 | 373 | 4647090 | ||
BC022334.1 | - | 3 | 355 | 4703567 | ||
BG034032.1 | - | 3 | 295 | |||
BQ431723.1 | - | 3 | 408 | |||
EU794640.1 | - | 3 | 395 |