Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037162.1 | 11 | 2863 | |||
GenBank mRNA | AK297289.1 | - | 10 | 2000 | ||
AL096725.1 | - | 5 | 2188 | |||
AL833939.1 | - | 8 | 2978 | |||
BC098298.1 | - | 6 | 1798 | 40025661 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 12 | 2690 | ||
GenBank mRNA | BC025289.1 | - | 11 | 2124 |