Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002531.2 | NP_002522.2 | 1, 2 | 1081 | ||
XM_011528827.2 | XP_011527129.1 | 1, 2 | 1148 | |||
GenBank mRNA | AY429106.1 | - | 1, 2 | 710 | ||
X70070.1 | - | 1, 2 | 1082 |
Set Membership: |
G-protein coupled receptor |