Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173476.1 | NP_001166947.1 | 5 | 1389 | ||
XM_017003162.1 | XP_016858651.1 | 8 | 3166 | |||
GenBank mRNA | AK055387.1 | - | 5 | 3445 | ||
AK126500.1 | - | 11 | 3832 | |||
AK128689.1 | - | 9 | 5078 | |||
AK129604.1 | - | 4 | 1846 | |||
AK289531.1 | - | 12 | 3169 | |||
BC006346.2 | - | 5 | 1231 | |||
U57099.1 | - | 5 | 1219 |