Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006049.2 | NP_006040.1 | 3 | 359 | ||
GenBank mRNA | AF093593.1 | - | 3 | 282 | ||
AK024763.1 | - | 3 | 758 | |||
AK292051.1 | - | 2 | 207 | |||
AK312018.1 | - | 3 | 331 | |||
BC014315.1 | - | 2 | 227 | 3997264 | ||
CD685245.1 | - | 3 | 396 | |||
CN347010.1 | - | 3 | 281 | |||
DB037177.1 | - | 3 | 291 |