Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 2, 3 | 442 | ||
NM_001319103.1 | NP_001306032.1 | 1, 2 | 720 | |||
NM_001319104.1 | NP_001306033.1 | 1, 2 | 357 | |||
NM_001319105.1 | NP_001306034.1 | 2, 3 | 266 | |||
NM_001319106.1 | NP_001306035.1 | 1, 2 | 720 | |||
NM_002545.4 | NP_002536.1 | 1, 2 | 720 | |||
XM_006718846.2 | XP_006718909.1 | 2, 3 | 668 | |||
XM_011542856.2 | XP_011541158.1 | 3, 4 | 978 | |||
GenBank mRNA | AK289695.1 | - | 1, 2 | 348 | ||
AK299528.1 | - | 2, 3 | 193 | |||
AK299908.1 | - | 2, 3 | 229 | |||
AK314077.1 | - | 1, 2 | 720 | |||
AK316273.1 | - | 1, 2 | 144 | |||
AK316491.1 | - | 2, 3 | 266 | |||
BC074742.2 | - | 1, 2 | 202 | 30915458 | ||
BC074773.2 | - | 1, 2 | 202 | 30915200 | ||
BC117254.1 | - | 1, 2 | 198 | 40125805 | ||
BC126251.1 | - | 1, 2 | 198 | 8991967 | ||
BC143945.1 | - | 1, 2 | 198 | 9052463 | ||
BC143946.1 | - | 1, 2 | 198 | 9052464 | ||
BX537377.1 | - | 2, 3 | 290 | |||
DC332150.1 | - | 442 | ||||
EU562295.1 | - | 1, 2 | 754 | |||
EU562296.1 | - | 2, 3 | 232 | |||
EU562297.1 | - | 2, 3 | 169 | |||
EU562298.1 | - | 1, 2 | 147 | |||
EU562299.1 | - | 1, 2 | 59 | |||
L34774.1 | - | 1, 2 | 225 |