Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098527.2 | NP_001091997.2 | 5 | 2162 | ||
NM_001159545.1 | NP_001153017.1 | 4 | 1204 | |||
NM_001159546.1 | NP_001153018.1 | 4 | 1233 | |||
NM_006605.3 | NP_006596.2 | 2 | 1248 | |||
XM_011529826.1 | XP_011528128.1 | 5 | 2366 | |||
XM_011529827.2 | XP_011528129.1 | 6 | 1541 | |||
XM_011529828.1 | XP_011528130.1 | 6 | 2276 | |||
XM_011529830.2 | XP_011528132.1 | 6 | 1626 | |||
XM_011529832.1 | XP_011528134.1 | 4 | 2252 | |||
XM_017028535.1 | XP_016884024.1 | 5 | 1371 | |||
XM_017028536.1 | XP_016884025.1 | 5 | 1420 | |||
GenBank mRNA | AJ010231.1 | - | 2 | 1248 | ||
AK055999.1 | - | 5 | 2162 | |||
DB549304.1 | - | 1 | 209 |