Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315501.1 | NP_001302430.1 | 7 | 1164 | ||
NM_001315502.1 | NP_001302431.1 | 5 | 654 | |||
NM_002555.5 | NP_002546.3 | 7 | 948 | |||
NM_183233.2 | NP_899056.2 | 7 | 951 | |||
XM_011520141.2 | XP_011518443.2 | 7 | 1107 | |||
XM_011520142.2 | XP_011518444.2 | 7 | 1106 | |||
GenBank mRNA | AB012083.1 | - | 7 | 927 | ||
AF030302.1 | - | 7 | 1164 | |||
AF037064.1 | - | 7 | 935 | |||
AF059663.1 | - | 7 | 927 | |||
AL521035.3 | - | 5 | 651 | |||
AL832892.1 | - | 2 | 673 | |||
BC015571.2 | - | 7 | 951 | 3636077 |