Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193513.1 | NP_001180442.1 | 15 | 2284 | ||
NM_001193514.1 | NP_001180443.1 | 13 | 2095 | |||
NM_001193515.1 | NP_001180444.1 | 13 | 2077 | |||
NM_017964.3 | NP_060434.2 | 14 | 2164 | |||
XM_006712048.3 | XP_006712111.1 | 11 | 1946 | |||
XM_011532959.2 | XP_011531261.1 | 14 | 2192 | |||
XM_011532960.2 | XP_011531262.1 | 14 | 2420 | |||
XM_011532961.2 | XP_011531263.1 | 13 | 2098 | |||
XM_011532962.2 | XP_011531264.1 | 12 | 2249 | |||
XM_017004462.1 | XP_016859951.1 | 12 | 2011 | |||
GenBank mRNA | AF173387.1 | - | 1143 | |||
AK055663.1 | - | 14 | 2141 | |||
AK096672.1 | - | 13 | 2124 | |||
AK127716.1 | - | 10 | 1898 | |||
AL832755.1 | - | 11 | 1921 | |||
BC032525.1 | - | 5 | 1514 | |||
BC066903.1 | - | 15 | 2261 | 4801303 |