Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002558.3 | NP_002549.1 | 4 | 830 | ||
XM_006721529.2 | XP_006721592.1 | 4 | 670 | |||
XM_011523896.2 | XP_011522198.1 | 5 | 1222 | |||
XM_011523897.2 | XP_011522199.1 | 4 | 670 | |||
XM_011523898.2 | XP_011522200.1 | 4 | 1334 | |||
XM_011523899.2 | XP_011522201.1 | 4 | 1334 | |||
XM_011523900.2 | XP_011522202.1 | 4 | 1334 | |||
GenBank mRNA | AF020498.1 | - | 4 | 665 | ||
AK313264.1 | - | 4 | 629 | |||
BC027949.1 | - | 4 | 616 | |||
BC044657.1 | - | 4 | 605 | 5756349 | ||
U45448.1 | - | 4 | 585 | |||
X83688.1 | - | 4 | 562 |