Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164692.2 | NP_001158164.1 | 2 | 175 | ||
NM_019839.4 | NP_062813.2 | 2 | 119 | |||
GenBank mRNA | AB032558.1 | - | 2 | 85 | ||
AF277230.1 | - | 1 | 1715 | |||
AK304169.1 | - | 2 | 111 | |||
BC038437.1 | - | 2 | 155 | |||
BC119674.2 | - | 1 | 309 | |||
BC127643.1 | - | 1 | 312 | |||
DC408599.1 | - | 2 | 119 |
Set Membership: |
G-protein coupled receptor |