Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002564.3 | NP_002555.3 | 3 | 429 | ||
NM_176071.2 | NP_788085.2 | 3 | 494 | |||
NM_176072.2 | NP_788086.2 | 3 | 563 | |||
XM_005274019.4 | XP_005274076.1 | 3 | 381 | |||
XM_005274020.4 | XP_005274077.1 | 3 | 1968 | |||
XM_005274021.4 | XP_005274078.1 | 3 | 1562 | |||
XM_011545074.2 | XP_011543376.1 | 3 | 316 | |||
XM_017017839.1 | XP_016873328.1 | 4 | 441 | |||
GenBank mRNA | AK313942.1 | - | 3 | 390 | ||
AY136753.1 | - | 1 | 95 | |||
BC012104.1 | - | 3 | 562 | 4561430 | ||
BC028135.1 | - | 3 | 494 | 5215817 | ||
U07225.1 | - | 3 | 340 |
Set Membership: |
G-protein coupled receptor |