Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024036.4 | NP_076941.2 | 1 | 1642 | ||
XM_005274239.3 | XP_005274296.1 | 2 | 1522 | |||
GenBank mRNA | AB209005.1 | - | 1 | 1263 | ||
BC000207.1 | - | 1 | 379 | |||
BC007718.2 | - | 1 | 1522 | |||
BC014040.2 | - | 1 | 386 | |||
BC015581.2 | - | 1 | 1545 | 4637053 | ||
BC027475.1 | - | 1 | 981 | |||
BC094813.1 | - | 1 | 720 |