Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPDPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024299.2 | NP_077275.1 | 4 | 318 | ||
GenBank mRNA | AK024699.1 | - | 4 | 304 | ||
BC002531.2 | - | 4 | 318 | 3140372 | ||
BC056416.1 | - | 4 | 372 | 6138381 |