Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024015.4 | NP_076920.1 | 2 | 718 | ||
GenBank mRNA | AL137449.1 | - | 2 | 641 | ||
BC049204.1 | - | 2 | 718 | 5533346 |