Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016417.2 | NP_057501.2 | 2 | 873 | ||
GenBank mRNA | AB223038.1 | - | 2 | 812 | ||
AF113691.1 | - | 443 | ||||
BC023528.2 | - | 2 | 823 | 3613760 | ||
BC047680.1 | - | 2 | 819 | 6066312 | ||
BC105275.1 | - | 1 | 350 | |||
BI834047.1 | - | 2 | 621 | |||
BX248075.1 | - | 2 | 826 |