Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020064.3 | NP_064448.1 | 3 | 1700 | ||
GenBank mRNA | AF325688.1 | - | 3 | 1523 | ||
AK054797.1 | - | 3 | 1700 |