Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021175.3 | NP_066998.1 | 3 | 551 | ||
GenBank mRNA | AF131292.1 | - | 3 | 365 | ||
AF147355.1 | - | 1 | 71 | |||
AF309489.1 | - | 3 | 390 | |||
AJ277280.1 | - | 3 | 357 | |||
AK096110.1 | - | 1 | 2595 | |||
AY358669.1 | - | 3 | 359 | |||
BC020612.1 | - | 3 | 369 | 4715540 |