Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021221.2 | NP_067044.2 | 3 | 404 | ||
GenBank mRNA | AJ245417.1 | - | 2 | 1123 | ||
AJ315545.1 | - | 3 | 404 | |||
AK289364.1 | - | 3 | 1188 | |||
CF264683.1 | - | 4 | 437 | |||
HE864450.1 | - | 3 | 424 | |||
HE864451.1 | - | 2 | 571 |
Gene Symbol: CSNK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HE864455.1 | - | 4 | 296 |