Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022006.1 | NP_071289.1 | 5 | 293 | ||
GenBank mRNA | AI929519.1 | - | 5 | 284 | ||
AK057825.1 | - | 5 | 268 | |||
BC018619.1 | - | 5 | 277 | 3626060 |