Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024036.4 | NP_076941.2 | 1 | 1678 | ||
XM_005274239.3 | XP_005274296.1 | 2 | 1558 | |||
GenBank mRNA | AB209005.1 | - | 1 | 1299 | ||
BC000207.1 | - | 1 | 415 | |||
BC014040.2 | - | 1 | 422 | |||
BC015581.2 | - | 1 | 1581 | 4637053 | ||
BC027475.1 | - | 1 | 1017 | |||
BC094813.1 | - | 1 | 756 |