Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024509.1 | NP_078785.1 | 2 | 1834 | ||
XM_017027302.1 | XP_016882791.1 | 1 | 1399 | |||
GenBank mRNA | AK172754.1 | - | 1 | 1382 | ||
AK313579.1 | - | 2 | 1498 | |||
AY358127.1 | - | 2 | 2114 | |||
BC003578.1 | - | 2 | 1834 | 3537259 |