Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177591.1 | NP_001171062.1 | 3 | 205 | ||
NM_002717.3 | NP_002708.1 | 3 | 473 | |||
XM_005273559.1 | XP_005273616.1 | 3 | 438 | |||
XM_005273560.1 | XP_005273617.1 | 4 | 521 | |||
XM_011544565.1 | XP_011542867.1 | 3 | 829 | |||
XM_011544566.1 | XP_011542868.1 | 3 | 460 | |||
XM_011544567.2 | XP_011542869.1 | 3 | 441 | |||
XM_011544568.1 | XP_011542870.1 | 3 | 449 | |||
XM_011544569.1 | XP_011542871.1 | 3 | 181 | |||
GenBank mRNA | AK303981.1 | - | 3 | 207 | ||
AK314823.1 | - | 3 | 436 | |||
AY189686.1 | - | 3 | 170 | |||
BC041071.1 | - | 3 | 423 | 5484483 | ||
BX440554.2 | - | 3 | 447 | |||
M64929.1 | - | 3 | 249 |