Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290212.1 | NP_001277141.1 | 3 | 568 | ||
NM_031945.4 | NP_114151.3 | 2 | 334 | |||
NR_110866.1 | 2 | 334 | ||||
GenBank mRNA | AB593081.1 | - | 3 | 541 | ||
AB593082.1 | - | 2 | 329 | |||
AF325213.1 | - | 2 | 290 | |||
AK096926.1 | - | 2 | 316 | |||
BC021923.1 | - | 2 | 303 | 4876887 | ||
BC032802.1 | - | 2 | 305 | 5191718 | ||
BG760374.1 | - | 2 | 303 | |||
BM710905.1 | - | 2 | 336 | |||
DA912496.1 | - | 2 | 316 |