Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303503.1 | NP_001290432.1 | 2, 3 | 289 | ||
NM_001303504.1 | NP_001290433.1 | 2, 3 | 283 | |||
NM_002720.2 | NP_002711.1 | 2, 3 | 283 | |||
XM_006721061.3 | XP_006721124.1 | 2, 3 | 270 | |||
GenBank mRNA | AF097996.1 | - | 2, 3 | 205 | ||
BC001416.2 | - | 2, 3 | 236 | 3140324 | ||
BQ220878.1 | - | 2, 3 | 193 | |||
BT019746.1 | - | 1, 2 | 101 | |||
X70218.1 | - | 2, 3 | 236 |