Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318015.1 | NP_001304944.1 | 3 | 452 | ||
NM_001318016.1 | NP_001304945.1 | 3 | 287 | |||
NM_001318020.1 | NP_001304949.1 | 3 | 287 | |||
NM_001318023.1 | NP_001304952.1 | 3 | 287 | |||
NM_032799.4 | NP_116188.2 | 3 | 287 | |||
XM_011519116.2 | XP_011517418.1 | 3 | 461 | |||
GenBank mRNA | AK027430.1 | - | 3 | 280 | ||
AK075332.1 | - | 3 | 280 | |||
AK092843.1 | - | 2 | 1704 | |||
AK315746.1 | - | 3 | 283 | |||
BC009280.2 | - | 3 | 264 | 4302257 | ||
BC048251.1 | - | 3 | 433 | 5418875 | ||
BG767826.1 | - | 3 | 261 | |||
BM043458.1 | - | 3 | 255 | |||
BM745076.1 | - | 3 | 279 | |||
BU506687.1 | - | 3 | 288 |