Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052863.2 | NP_443095.2 | 3 | 354 | ||
GenBank mRNA | AF086152.1 | - | 3 | 350 | ||
AF436839.1 | - | 3 | 350 | |||
AY040564.1 | - | 3 | 350 | |||
AY359064.1 | - | 3 | 407 | |||
BC029176.1 | - | 3 | 344 | 5180304 | ||
BC072673.1 | - | 3 | 350 | 5753142 |