Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080819.4 | NP_543009.2 | 1 | 742 | ||
GenBank mRNA | AF411107.1 | - | 1 | 249 | ||
AK128807.1 | - | 1 | 742 | |||
AK313038.1 | - | 1 | 317 | |||
AY359107.1 | - | 1 | 543 | |||
BC057778.1 | - | 1 | 666 | 30343110 | ||
BC069343.1 | - | 1 | 249 | 7262307 | ||
BC069813.1 | - | 1 | 249 | 7262319 | ||
JF810885.1 | - | 1 | 249 |
Set Membership: |
G-protein coupled receptor |