Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138689.2 | NP_619634.1 | 2 | 538 | ||
GenBank mRNA | AF318364.1 | - | 1 | 539 | ||
BC014522.2 | - | 2 | 390 | 3831227 | ||
BC071884.1 | - | 1 | 293 | |||
BC094817.1 | - | 2 | 421 | 5466876 | ||
BT009914.1 | - | 2 | 270 | |||
X91195.1 | - | 2 | 522 |