Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP30L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131062.1 | NP_001124534.1 | 1 | 830 | ||
NM_001131063.1 | NP_001124535.1 | 1 | 830 | |||
NM_024632.5 | NP_078908.1 | 1 | 830 | |||
NR_024084.1 | 1 | 830 | ||||
GenBank mRNA | AI144341.1 | - | 1 | 242 | ||
AI199517.1 | - | 1 | 242 | |||
AI436556.1 | - | 1 | 242 | |||
AK021588.1 | - | 1 | 830 | |||
AY341060.1 | - | 1 | 773 | |||
AY846876.1 | - | 1 | 182 | |||
BC009829.2 | - | 1 | 764 | 3937610 |