Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144037.1 | NP_001137509.1 | 5, 6 | 972 | ||
NM_001318755.1 | NP_001305684.1 | 5, 6 | 972 | |||
NM_001318757.1 | NP_001305686.1 | 4, 5 | 681 | |||
NM_032780.3 | NP_116169.2 | 5, 6 | 972 | |||
XM_005271703.1 | XP_005271760.1 | 5, 6 | 881 | |||
XM_006718926.2 | XP_006718989.1 | 5, 6 | 856 | |||
XM_011543037.2 | XP_011541339.1 | 5, 6 | 856 | |||
XM_017018422.1 | XP_016873911.1 | 5, 6 | 881 | |||
XM_017018423.1 | XP_016873912.1 | 5, 6 | 881 | |||
XM_017018424.1 | XP_016873913.1 | 5, 6 | 762 | |||
XM_017018425.1 | XP_016873914.1 | 5, 6 | 785 | |||
XM_017018427.1 | XP_016873916.1 | 5, 6 | 762 | |||
XM_017018428.1 | XP_016873917.1 | 5, 6 | 785 | |||
XM_017018429.1 | XP_016873918.1 | 5, 6 | 762 | |||
GenBank mRNA | AK027305.1 | - | 5, 6 | 861 | ||
AK225415.1 | - | 5, 6 | 858 | |||
AK293670.1 | - | 4, 5 | 577 | |||
BC042896.1 | - | 5, 6 | 851 | 5168151 |