Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP9-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033191.2 | NP_149461.2 | 1 | 927 | ||
GenBank mRNA | AJ406948.1 | - | 1 | 927 |