Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080819.4 | NP_543009.2 | 2 | 1223 | ||
NR_045511.2 | 2 | 530 | ||||
GenBank mRNA | AF411107.1 | - | 2 | 730 | ||
AK128807.1 | - | 2 | 1223 | |||
AK307281.1 | - | 2 | 530 | |||
AK313038.1 | - | 2 | 798 | |||
AY359107.1 | - | 2 | 1024 | |||
BC057778.1 | - | 2 | 1147 | 30343110 | ||
BC069343.1 | - | 2 | 730 | 7262307 | ||
BC069813.1 | - | 2 | 730 | 7262319 | ||
JF810885.1 | - | 2 | 730 |
Set Membership: |
G-protein coupled receptor |