Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138689.2 | NP_619634.1 | 2 | 543 | ||
GenBank mRNA | AF318364.1 | - | 1 | 544 | ||
BC014522.2 | - | 2 | 395 | 3831227 | ||
BC071884.1 | - | 1 | 298 | |||
BC094817.1 | - | 2 | 426 | 5466876 | ||
BT009914.1 | - | 2 | 275 | |||
X91195.1 | - | 2 | 527 |