Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030891.4 | NP_112153.1 | 2 | 742 | ||
GenBank mRNA | AB058646.1 | - | 1 | 435 | ||
AY061857.1 | - | 1 | 435 | |||
AY358120.1 | - | 2 | 704 | |||
BC064321.1 | - | 2 | 742 | |||
BC119648.2 | - | 1 | 532 | 40115796 | ||
BC119649.1 | - | 1 | 535 | 40115797 |