Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 1337 | |||
NM_001290139.1 | NP_001277068.1 | 1407 | ||||
NM_001323636.1 | NP_001310565.1 | 1362 | ||||
NM_032304.3 | NP_115680.1 | 1368 | ||||
GenBank mRNA | BC004353.1 | - | 1343 | 3351990 | ||
BC015008.1 | - | 1296 | 4862387 | |||
BC033796.1 | - | 1697 | 5173915 | |||
BC036747.1 | - | 1697 |