Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130900.2 | NP_570970.2 | 3 | 507 | ||
XM_011535486.2 | XP_011533788.1 | 3 | 571 | |||
GenBank mRNA | AY039682.1 | - | 3 | 446 | ||
BC131600.1 | - | 3 | 439 |