Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053056.2 | NP_444284.1 | 3 | 733 | ||
GenBank mRNA | AK313136.1 | - | 3 | 682 | ||
BC000076.2 | - | 3 | 667 | 3508088 | ||
BC001501.2 | - | 3 | 667 | 3507598 | ||
BC014078.2 | - | 3 | 667 | 4124540 | ||
BC023620.2 | - | 3 | 660 | 4650919 | ||
BC025302.1 | - | 3 | 669 | 5457963 | ||
BT019844.1 | - | 3 | 524 | |||
BT019845.1 | - | 3 | 524 | |||
CR536538.1 | - | 3 | 524 | |||
CR542099.1 | - | 3 | 524 | |||
M64349.1 | - | 3 | 668 | |||
M73554.1 | - | 3 | 665 | |||
M74092.1 | - | 3 | 673 | |||
X59798.1 | - | 3 | 671 |