Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178130.2 | NP_001171601.1 | 2 | 647 | ||
NM_001178131.2 | NP_001171602.1 | 2 | 647 | |||
NM_001963.5 | NP_001954.2 | 2 | 647 | |||
XM_005262796.2 | XP_005262853.1 | 2 | 648 | |||
XM_011531707.1 | XP_011530009.1 | 3 | 297 | |||
XM_017007845.1 | XP_016863334.1 | 2 | 672 | |||
XM_017007846.1 | XP_016863335.1 | 2 | 672 | |||
XM_017007847.1 | XP_016863336.1 | 2 | 672 | |||
XM_017007848.1 | XP_016863337.1 | 2 | 672 | |||
XM_017007849.1 | XP_016863338.1 | 2 | 672 | |||
XM_017007850.1 | XP_016863339.1 | 2 | 672 | |||
XM_017007851.1 | XP_016863340.1 | 2 | 672 | |||
XM_017007852.1 | XP_016863341.1 | 2 | 648 | |||
XM_017007853.1 | XP_016863342.1 | 2 | 672 | |||
XM_017007854.1 | XP_016863343.1 | 2 | 672 | |||
XM_017007855.1 | XP_016863344.1 | 2 | 672 | |||
GenBank mRNA | AK299306.1 | - | 2 | 649 | ||
BC093731.1 | - | 2 | 290 | 7939576 | ||
BC113461.1 | - | 2 | 290 | 8322513 | ||
BC143357.1 | - | 2 | 290 | |||
X04571.1 | - | 2 | 631 |