Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005225.2 | NP_005216.1 | 2 | 420 | ||
GenBank mRNA | BC050369.2 | - | 2 | 420 | 6025053 | |
BC058902.1 | - | 2 | 351 | 6055344 | ||
M96577.1 | - | 2 | 415 | |||
S49592.1 | - | 2 | 397 |