Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256252.1 | NP_001243181.1 | 3 | 249 | ||
NM_001256253.1 | NP_001243182.1 | 3 | 249 | |||
NM_001256254.1 | NP_001243183.1 | 3 | 249 | |||
NM_001256255.1 | NP_001243184.1 | 3 | 249 | |||
NM_001974.4 | NP_001965.3 | 3 | 249 | |||
XM_011527794.1 | XP_011526096.1 | 3 | 249 | |||
GenBank mRNA | AK131562.1 | - | 3 | 219 | ||
AK290401.1 | - | 3 | 219 | |||
AK291518.1 | - | 3 | 219 | |||
AK297011.1 | - | 3 | 222 | |||
AK313495.1 | - | 3 | 219 | |||
BC059395.1 | - | 3 | 232 | 30349201 | ||
DA465986.1 | - | 3 | 249 | |||
X81479.1 | - | 3 | 227 |
Set Membership: |
G-protein coupled receptor |