Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 3 | 638 | ||
GenBank mRNA | AK291325.1 | - | 3 | 580 | ||
AK312657.1 | - | 3 | 586 | |||
BC064803.1 | - | 3 | 572 | 6046773 | ||
D26512.1 | - | 3 | 464 | |||
HE978355.1 | - | 3 | 587 | |||
U41078.1 | - | 3 | 353 | |||
X83535.1 | - | 3 | 472 | |||
X90925.1 | - | 3 | 465 | |||
Z48481.1 | - | 3 | 466 |