Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002513.2 | NP_002504.2 | 2 | 321 | ||
XM_005255332.4 | XP_005255389.1 | 2 | 352 | |||
XM_011522503.2 | XP_011520805.1 | 2 | 362 | |||
XM_011522504.2 | XP_011520806.1 | 2 | 357 | |||
GenBank mRNA | AK308663.1 | - | 2 | 122 | ||
BC000250.1 | - | 2 | 321 | 3357930 | ||
BG911570.1 | - | 2 | 145 | |||
U29656.1 | - | 2 | 164 |