Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177983.2 | NP_817092.1 | 1, 2 | 389 | ||
GenBank mRNA | AB209197.1 | - | 1, 2 | 261 | ||
AI879284.1 | - | 1, 2 | 339 | |||
AK127593.1 | - | 2 | 255 | |||
AK293141.1 | - | 2, 3 | 183 | |||
AK312455.1 | - | 1, 2 | 379 | |||
BC000057.2 | - | 1, 2 | 238 | 3505271 | ||
BC022061.2 | - | 1, 2 | 235 | 2988852 | ||
BT007454.1 | - | 1, 2 | 128 | |||
CR457059.1 | - | 1, 2 | 128 | |||
DA520893.1 | - | 1, 2 | 389 | |||
Y13936.1 | - | 1, 2 | 152 |