Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012084.3 | NP_036216.2 | 1 | 331 | ||
GenBank mRNA | AK313356.1 | - | 1 | 328 | ||
BC050732.2 | - | 1 | 331 | 6164919 |